Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258390.1 | NP_001245319.1 | 7 | 1109 | ||
NM_001258391.1 | NP_001245320.1 | 5 | 927 | |||
NM_001318799.1 | NP_001305728.1 | 6 | 719 | |||
NM_178834.4 | NP_849156.1 | 6 | 1085 | |||
XM_006718769.3 | XP_006718832.1 | 6 | 698 | |||
XM_006718770.3 | XP_006718833.1 | 5 | 755 | |||
GenBank mRNA | AK055539.1 | - | 6 | 1086 | ||
AK055654.1 | - | 6 | 873 | |||
AK293317.1 | - | 6 | 916 | |||
AK296231.1 | - | 5 | 715 | |||
AK298165.1 | - | 7 | 744 | |||
AY358554.1 | - | 7 | 934 | |||
BC025407.1 | - | 6 | 878 | 4820414 |