Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302269.1 | NP_001289198.1 | 4 | 1071 | ||
NM_001956.4 | NP_001947.1 | 5 | 1170 | |||
NR_126098.1 | 5 | 1079 | ||||
XM_017000512.1 | XP_016856001.1 | 3 | 963 | |||
GenBank mRNA | BC034393.1 | - | 5 | 1113 | 5185394 | |
M65199.1 | - | 5 | 1158 | |||
X55177.1 | - | 5 | 1146 |