Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2AG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004489.2 | NP_001004489.1 | 1 | 610 | ||
GenBank mRNA | BC151143.1 | - | 1 | 610 | 9020805 |