Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144881.1 | NP_001138353.1 | 7 | 1896 | ||
NM_001144882.1 | NP_001138354.1 | 7 | 1895 | |||
NM_001144883.1 | NP_001138355.1 | 7 | 1892 | |||
NM_153026.2 | NP_694571.2 | 7 | 1963 | |||
XM_011537946.1 | XP_011536248.1 | 7 | 1714 | |||
XM_011537947.2 | XP_011536249.1 | 7 | 1945 | |||
XM_017018838.1 | XP_016874327.1 | 7 | 1759 | |||
XM_017018839.1 | XP_016874328.1 | 7 | 1959 | |||
XM_017018840.1 | XP_016874329.1 | 7 | 1779 | |||
GenBank mRNA | AF399844.1 | - | 7 | 1869 | ||
AK056189.1 | - | 1 | 487 | |||
AK056499.1 | - | 7 | 1869 | |||
AK122877.1 | - | 7 | 1897 | |||
AK122881.1 | - | 7 | 1840 | |||
BC114939.1 | - | 7 | 1804 | 40084889 | ||
BC114940.1 | - | 7 | 1805 | 40084893 |