Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 1352 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 1133 | |||
GenBank mRNA | AY845858.1 | - | 3 | 1213 | ||
BC137413.1 | - | 3 | 1268 | 9021411 | ||
BC137414.1 | - | 3 | 1268 | 9021412 | ||
BC144617.1 | - | 3 | 1242 | |||
DB224774.1 | - | 1 | 195 |