Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204298.1 | NP_001191227.1 | 5 | 1029 | ||
NM_152437.2 | NP_689650.1 | 5 | 1081 | |||
GenBank mRNA | AK056034.1 | - | 5 | 988 | ||
AL834266.1 | - | 6 | 1882 | |||
BC017960.1 | - | 1 | 344 | 4608465 | ||
BC051696.1 | - | 5 | 1011 | 5276234 | ||
BC093848.1 | - | 1 | 92 | 7939693 | ||
BC101530.1 | - | 1 | 92 | 8069036 | ||
BX647762.1 | - | 6 | 1001 |