Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001660.2 | NP_001001660.2 | 2, 3 | 220 | ||
XM_005253319.4 | XP_005253376.1 | 2, 3 | 223 | |||
XM_005253320.4 | XP_005253377.1 | 3, 4 | 327 | |||
XM_017018850.1 | XP_016874339.1 | 2, 3 | 1641 | |||
GenBank mRNA | AK057730.1 | - | 2, 3 | 224 | ||
BC071769.1 | - | 2, 3 | 154 | 4246777 | ||
BC100276.1 | - | 2, 3 | 171 | 6178563 | ||
BG771497.1 | - | 2, 3 | 225 |