Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286531.1 | NP_001273460.1 | 9 | 897 | ||
NM_001286532.1 | NP_001273461.1 | 9 | 919 | |||
NM_001286533.1 | NP_001273462.1 | 10 | 1075 | |||
NM_001286534.1 | NP_001273463.1 | 10 | 995 | |||
NM_001286535.1 | NP_001273464.1 | 9 | 926 | |||
NM_001286536.1 | NP_001273465.1 | 7 | 719 | |||
NM_152442.3 | NP_689655.3 | 9 | 926 | |||
XM_005253849.4 | XP_005253906.1 | 7 | 605 | |||
XM_011537971.2 | XP_011536273.1 | 9 | 881 | |||
XM_011537972.2 | XP_011536274.1 | 9 | 754 | |||
XM_011537974.2 | XP_011536276.1 | 8 | 757 | |||
XM_017018877.1 | XP_016874366.1 | 8 | 670 | |||
XM_017018878.1 | XP_016874367.1 | 9 | 874 | |||
XM_017018879.1 | XP_016874368.1 | 8 | 810 | |||
XM_017018880.1 | XP_016874369.1 | 8 | 718 | |||
XM_017018881.1 | XP_016874370.1 | 8 | 810 | |||
XM_017018882.1 | XP_016874371.1 | 8 | 662 | |||
GenBank mRNA | AK058176.1 | - | 5 | 320 | ||
AK097665.1 | - | 10 | 995 | |||
AK124109.1 | - | 9 | 897 | |||
AK301826.1 | - | 9 | 887 | |||
AK302512.1 | - | 7 | 605 | |||
AY297459.1 | - | 9 | 802 | |||
BC047645.1 | - | 10 | 942 | 4828083 | ||
BC068031.1 | - | 9 | 802 | 30378632 |