Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128303.1 | NP_001121775.1 | 5 | 1592 | ||
NM_153218.2 | NP_694950.2 | 5 | 1428 | |||
XM_005266261.3 | XP_005266318.1 | 5 | 2051 | |||
XM_006719766.3 | XP_006719829.1 | 5 | 1209 | |||
XM_006719767.3 | XP_006719830.1 | 5 | 1428 | |||
XM_011534933.2 | XP_011533235.1 | 6 | 2246 | |||
XM_011534934.2 | XP_011533236.1 | 5 | 1365 | |||
XM_017020394.1 | XP_016875883.1 | 5 | 1485 | |||
GenBank mRNA | AK096044.1 | - | 5 | 1428 | ||
BC035749.1 | - | 5 | 1397 | 5750179 | ||
BX538036.1 | - | 1 | 163 |