Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194318.3 | NP_919299.3 | 12 | 1112 | ||
XM_006719768.3 | XP_006719831.1 | 13 | 1302 | |||
XM_011534936.1 | XP_011533238.1 | 12 | 1101 | |||
XM_011534938.2 | XP_011533240.1 | 11 | 950 | |||
XM_017020395.1 | XP_016875884.1 | 13 | 1205 | |||
GenBank mRNA | AB101481.1 | - | 12 | 1002 | ||
AK291273.1 | - | 12 | 1101 | |||
AY190526.1 | - | 12 | 1002 | |||
BC047395.1 | - | 10 | 1066 | |||
BC068595.1 | - | 12 | 1092 | 4837250 | ||
DA290408.1 | - | 5 | 409 |