Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304461.1 | NP_001291390.1 | 3 | 1004 | ||
NM_173850.3 | NP_776249.1 | 4 | 1577 | |||
XM_011536451.2 | XP_011534753.1 | 3 | 980 | |||
XM_011536452.2 | XP_011534754.1 | 3 | 974 | |||
XM_011536453.2 | XP_011534755.1 | 3 | 886 | |||
XM_011536454.2 | XP_011534756.1 | 3 | 965 | |||
XM_011536455.1 | XP_011534757.1 | 2 | 798 | |||
XM_017020989.1 | XP_016876478.1 | 3 | 975 | |||
XM_017020990.1 | XP_016876479.1 | 3 | 960 | |||
GenBank mRNA | AY177692.1 | - | 2 | 799 | ||
AY326420.1 | - | 3 | 1004 | |||
BC040857.1 | - | 4 | 1577 | 5527002 |