Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 1606 | ||
XM_005253334.2 | XP_005253391.1 | 8 | 1606 | |||
XM_011520599.2 | XP_011518901.1 | 8 | 1609 | |||
XM_011520600.2 | XP_011518902.1 | 7 | 1483 | |||
XM_011520603.2 | XP_011518905.1 | 8 | 1713 | |||
XM_017018955.1 | XP_016874444.1 | 8 | 2001 | |||
XM_017018956.1 | XP_016874445.1 | 7 | 1459 | |||
XM_017018957.1 | XP_016874446.1 | 7 | 1432 | |||
XM_017018958.1 | XP_016874447.1 | 7 | 1408 | |||
GenBank mRNA | AB209063.1 | - | 9 | 1782 | ||
AK293893.1 | - | 8 | 1630 | |||
BC073964.1 | - | 3 | 1454 | 5788132 | ||
BC157850.1 | - | 2 | 1512 | 9057069 | ||
U89277.1 | - | 8 | 1585 |