Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172702.1 | NP_001166173.1 | 9 | 821 | ||
NM_153811.2 | NP_722518.2 | 9 | 821 | |||
NR_033344.1 | 9 | 997 | ||||
XM_006720049.3 | XP_006720112.1 | 9 | 773 | |||
XM_006720050.1 | XP_006720113.1 | 5 | 264 | |||
XM_011536469.2 | XP_011534771.1 | 9 | 773 | |||
XM_017021020.1 | XP_016876509.1 | 9 | 773 | |||
XM_017021021.1 | XP_016876510.1 | 9 | 773 | |||
XM_017021022.1 | XP_016876511.1 | 9 | 1086 | |||
XM_017021023.1 | XP_016876512.1 | 9 | 773 | |||
XM_017021025.1 | XP_016876514.1 | 9 | 1064 | |||
XM_017021026.1 | XP_016876515.1 | 9 | 1078 | |||
XM_017021027.1 | XP_016876516.1 | 7 | 442 | |||
XM_017021028.1 | XP_016876517.1 | 5 | 264 | |||
GenBank mRNA | AF070578.1 | - | 10 | 909 | ||
AF543422.1 | - | 9 | 773 | |||
AK303839.1 | - | 9 | 800 | |||
BC110378.1 | - | 9 | 682 | 5737780 | ||
BX248072.1 | - | 9 | 642 | |||
BX648332.1 | - | 9 | 695 |