Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172702.1 | NP_001166173.1 | 12 | 1018 | ||
NM_153811.2 | NP_722518.2 | 12 | 1018 | |||
NR_033344.1 | 12 | 1194 | ||||
XM_006720049.3 | XP_006720112.1 | 12 | 970 | |||
XM_006720050.1 | XP_006720113.1 | 8 | 461 | |||
XM_011536469.2 | XP_011534771.1 | 12 | 970 | |||
XM_017021020.1 | XP_016876509.1 | 12 | 970 | |||
XM_017021021.1 | XP_016876510.1 | 11 | 916 | |||
XM_017021022.1 | XP_016876511.1 | 12 | 1283 | |||
XM_017021023.1 | XP_016876512.1 | 12 | 970 | |||
XM_017021024.1 | XP_016876513.1 | 10 | 850 | |||
XM_017021025.1 | XP_016876514.1 | 12 | 1261 | |||
XM_017021026.1 | XP_016876515.1 | 12 | 1275 | |||
XM_017021027.1 | XP_016876516.1 | 10 | 639 | |||
XM_017021028.1 | XP_016876517.1 | 8 | 461 | |||
GenBank mRNA | AF070578.1 | - | 13 | 1106 | ||
AF543422.1 | - | 12 | 970 | |||
AK303839.1 | - | 12 | 997 | |||
BC110378.1 | - | 12 | 879 | 5737780 | ||
BX248072.1 | - | 12 | 839 | |||
BX648332.1 | - | 12 | 892 |