Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 685 | ||
XM_005253334.2 | XP_005253391.1 | 6 | 685 | |||
XM_011520599.2 | XP_011518901.1 | 6 | 688 | |||
XM_011520600.2 | XP_011518902.1 | 5 | 562 | |||
XM_011520603.2 | XP_011518905.1 | 6 | 792 | |||
XM_017018955.1 | XP_016874444.1 | 6 | 1080 | |||
XM_017018956.1 | XP_016874445.1 | 5 | 538 | |||
XM_017018957.1 | XP_016874446.1 | 5 | 511 | |||
XM_017018958.1 | XP_016874447.1 | 5 | 487 | |||
GenBank mRNA | AB209063.1 | - | 7 | 861 | ||
AK293893.1 | - | 6 | 709 | |||
BC073964.1 | - | 3 | 539 | 5788132 | ||
BC157850.1 | - | 2 | 591 | 9057069 | ||
CN388283.1 | - | 2 | 590 | |||
CN388316.1 | - | 1 | 85 | |||
U89277.1 | - | 6 | 664 |