Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001872.3 | NP_001001872.2 | 3 | 1680 | ||
NM_001320173.1 | NP_001307102.1 | 3 | 1680 | |||
XM_011536471.2 | XP_011534773.1 | 3 | 1680 | |||
XM_017021029.1 | XP_016876518.1 | 3 | 1537 | |||
XM_017021030.1 | XP_016876519.1 | 3 | 1680 | |||
GenBank mRNA | AK292513.1 | - | 3 | 1654 | ||
AK292605.1 | - | 3 | 1597 | |||
AK294599.1 | - | 2 | 891 | |||
BC013707.1 | - | 2 | 977 | |||
BC062772.1 | - | 2 | 301 | |||
BC146861.1 | - | 3 | 1531 | 9056789 | ||
BX161411.1 | - | 3 | 1570 | |||
BX161448.1 | - | 3 | 1624 | |||
BX649130.1 | - | 2 | 1176 |