Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194287.2 | NP_919263.2 | 6, 7 | 762 | ||
XM_011536479.1 | XP_011534781.1 | 6, 7 | 1130 | |||
XM_011536483.1 | XP_011534785.1 | 6, 7 | 1130 | |||
XM_011536487.1 | XP_011534789.1 | 6, 7 | 1130 | |||
XM_017021032.1 | XP_016876521.1 | 6, 7 | 1028 | |||
XM_017021033.1 | XP_016876522.1 | 6, 7 | 1130 | |||
XM_017021034.1 | XP_016876523.1 | 6, 7 | 774 | |||
XM_017021035.1 | XP_016876524.1 | 6, 7 | 925 | |||
XM_017021037.1 | XP_016876526.1 | 6, 7 | 1832 | |||
XM_017021038.1 | XP_016876527.1 | 6, 7 | 666 | |||
XM_017021039.1 | XP_016876528.1 | 5, 6 | 588 | |||
XM_017021040.1 | XP_016876529.1 | 6, 7 | 587 | |||
GenBank mRNA | AK058106.1 | - | 5, 6 | 675 | ||
AK125722.1 | - | 7, 8 | 985 | |||
BC062799.1 | - | 7, 8 | 1322 | |||
BC121159.2 | - | 6, 7 | 610 | 40121413 | ||
BC121160.2 | - | 6, 7 | 610 | 40121414 |