Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 435 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 436 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 511 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 145 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 461 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 561 | |||
NM_152449.3 | NP_689662.2 | 6 | 626 | |||
XM_006720397.3 | XP_006720460.2 | 2 | 159 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 219 | |||
XM_011521242.2 | XP_011519544.1 | 2 | 737 | |||
XM_011521243.2 | XP_011519545.1 | 2 | 737 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 788 | |||
XM_011521245.2 | XP_011519547.1 | 2 | 159 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 652 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 973 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 617 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 814 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 828 | |||
GenBank mRNA | AK124852.1 | - | 4 | 764 | ||
AK290296.1 | - | 3 | 421 | |||
BC041097.1 | - | 6 | 603 | 5240362 | ||
BC084545.1 | - | 3 | 403 | 4590701 | ||
BG420645.1 | - | 3 | 391 | |||
CB989579.1 | - | 4 | 452 | |||
DA405564.1 | - | 5 | 504 | |||
DA594971.1 | - | 2 | 119 | |||
DB044672.1 | - | 5 | 511 | |||
HY131530.1 | - | 3 | 172 |