Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4477
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
6
956
XM_011535514.2
XP_011533816.1
6
1591
GenBank mRNA
AF174496.1
-
5
786
AF328730.1
-
1
664
AF397403.1
-
4
701
AJ420488.1
-
5
916
AK024888.1
-
1
954
AK026650.1
-
1
956
AK055058.1
-
6
956
AK058051.1
-
6
956
AK058163.1
-
5
774
AK098510.1
-
6
956
AK098518.1
-
6
816
AK098587.1
-
3
383
AK098623.1
-
6
956
AK127384.1
-
6
956
AK222515.1
-
1
956
AK222519.1
-
1
605
AK222523.1
-
1
956
AK222551.1
-
1
956
AK222982.1
-
1
956
AK223030.1
-
1
953
AK223042.1
-
1
811
AK223046.1
-
1
959
AK289374.1
-
6
956
AK289442.1
-
6
894
AK290691.1
-
6
956
AK292639.1
-
6
1272
AK297878.1
-
7
754
AK300922.1
-
6
817
AK304214.1
-
7
916
AK315827.1
-
6
947
AY043301.1
-
5
1303
BC008587.1
-
1
942
3447654
BC009733.1
-
1
906
3877516
BC009875.2
-
1
946
3948601
BC010735.1
-
1
935
3898364
BC012891.1
-
1
934
3858483
BC014224.2
-
1
945
4335756
BC014377.1
-
1
368
BC014892.1
-
1
252
BC018150.1
-
1
942
3851939
BC018641.2
-
1
942
3660140
BC018850.2
-
1
945
BC019050.2
-
1
908
BC020477.2
-
1
605
BC021686.1
-
1
961
3930863
BC022412.1
-
1
257
BC028674.1
-
1
960
4107346
BC029337.1
-
1
930
BC029343.1
-
1
928
BC029997.1
-
1
943
BC035877.1
-
1
957
BC038339.1
-
1
942
3461588
BC057391.1
-
1
911
4339606
BC063511.1
-
1
379
BC065761.1
-
1
261
BC066893.1
-
1
956
4818863
BC071619.1
-
2
872
4448926
BC071727.1
-
1
911
4514479
BC071741.1
-
1
936
3885539
BC071841.1
-
1
931
4587981
BC072385.1
-
1
943
5585092
BC082268.1
-
1
935
6729595
BC094687.1
-
2
848
30366186
BC111051.1
-
1
946
5585079
EF219140.1
-
6
1116
EF362804.1
-
6
1186
M27364.1
-
2
194
M29548.1
-
3
488
X03558.1
-
6
946
X16869.1
-
5
908
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_175838, BC111707, L10339, X63561, BC128723, and others.This siRNA also targets the following transcripts in Mouse: NM_010106, AK213386, AK212450, AK197803, AK167873, and others.
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline