Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018100.4 | NP_001018110.1 | 6 | 700 | ||
NM_152451.7 | NP_689664.3 | 6 | 700 | |||
GenBank mRNA | AK055535.1 | - | 6 | 663 | ||
AL833142.1 | - | 5 | 481 | |||
AY207459.1 | - | 6 | 571 | |||
AY237639.1 | - | 6 | 571 | |||
BC101645.1 | - | 6 | 700 | 8069151 | ||
BC112148.1 | - | 6 | 698 | 8327616 | ||
FJ970029.1 | - | 6 | 576 |
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018090.6 | NP_001018100.1 | 6 | 700 | ||
NM_001018091.6 | NP_001018101.1 | 6 | 700 | |||
NM_001285900.3 | NP_001272829.1 | 6 | 700 | |||
NR_104367.2 | 6 | 700 | ||||
NR_104368.2 | 6 | 700 | ||||
NR_104369.2 | 6 | 700 | ||||
NR_104370.2 | 6 | 700 | ||||
NR_104371.3 | 6 | 700 | ||||
GenBank mRNA | AY207007.1 | - | 6 | 571 | ||
AY208913.1 | - | 6 | 571 | |||
AY331564.1 | - | 6 | 571 | |||
AY333779.1 | - | 6 | 571 | |||
AY334560.1 | - | 6 | 571 | |||
AY334561.1 | - | 6 | 571 | |||
AY334562.1 | - | 5 | 478 | |||
AY341345.1 | - | 4 | 364 | |||
AY353057.1 | - | 6 | 571 | |||
AY353058.1 | - | 6 | 571 | |||
AY353060.1 | - | 5 | 478 | |||
JF419331.1 | - | 6 | 590 |