Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4478
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
4
587
XM_011535514.2
XP_011533816.1
4
1222
GenBank mRNA
AF328730.1
-
1
295
AF397403.1
-
2
332
AJ420488.1
-
3
546
AK024888.1
-
1
585
AK026650.1
-
1
587
AK055058.1
-
4
587
AK058051.1
-
4
587
AK098510.1
-
4
587
AK098518.1
-
4
447
AK098587.1
-
1
15
AK098623.1
-
4
587
AK127384.1
-
4
587
AK222515.1
-
1
587
AK222519.1
-
1
236
AK222523.1
-
1
587
AK222551.1
-
1
587
AK223030.1
-
1
584
AK223042.1
-
1
442
AK223046.1
-
1
590
AK289374.1
-
4
587
AK289442.1
-
4
525
AK290691.1
-
4
587
AK292639.1
-
4
903
AK300922.1
-
4
448
AK304214.1
-
5
547
AK315827.1
-
4
578
AY043301.1
-
3
934
BC008587.1
-
1
573
3447654
BC009733.1
-
1
537
3877516
BC009875.2
-
1
577
3948601
BC010735.1
-
1
566
3898364
BC012891.1
-
1
565
3858483
BC014224.2
-
1
576
4335756
BC018150.1
-
1
573
3851939
BC018641.2
-
1
573
3660140
BC018850.2
-
1
576
BC019050.2
-
1
539
BC020477.2
-
1
236
BC021686.1
-
1
592
3930863
BC028674.1
-
1
591
4107346
BC029337.1
-
1
561
BC029343.1
-
1
559
BC029997.1
-
1
574
BC035877.1
-
1
588
BC038339.1
-
1
573
3461588
BC057391.1
-
1
542
4339606
BC066893.1
-
1
587
4818863
BC071619.1
-
2
503
4448926
BC071727.1
-
1
542
4514479
BC071741.1
-
1
567
3885539
BC071841.1
-
1
562
4587981
BC072385.1
-
1
574
5585092
BC082268.1
-
1
566
6729595
BC094687.1
-
2
479
30366186
BC111051.1
-
1
577
5585079
DQ867534.1
-
1
273
EF219140.1
-
4
747
EF362804.1
-
4
817
M29548.1
-
1
119
X03558.1
-
4
577
X16869.1
-
3
539
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline