Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138573.3 | NP_612640.1 | 4 | 395 | ||
XM_017021937.1 | XP_016877426.1 | 8 | 906 | |||
XM_017021938.1 | XP_016877427.1 | 7 | 3113 | |||
XM_017021939.1 | XP_016877428.1 | 7 | 675 | |||
XM_017021940.1 | XP_016877429.1 | 6 | 3451 | |||
XM_017021941.1 | XP_016877430.1 | 4 | 414 | |||
XM_017021942.1 | XP_016877431.1 | 4 | 432 | |||
XM_017021943.1 | XP_016877432.1 | 4 | 505 | |||
XM_017021944.1 | XP_016877433.1 | 7 | 3498 | |||
XM_017021945.1 | XP_016877434.1 | 7 | 3490 | |||
XM_017021946.1 | XP_016877435.1 | 7 | 3490 | |||
XM_017021947.1 | XP_016877436.1 | 7 | 3490 | |||
XM_017021948.1 | XP_016877437.1 | 5 | 3223 | |||
GenBank mRNA | AK307362.1 | - | 4 | 373 | ||
AK311204.1 | - | 9 | 727 | |||
AK311507.1 | - | 5 | 497 | |||
AM392364.1 | - | 2 | 213 | |||
BC017568.1 | - | 4 | 399 | 4071605 | ||
BX538100.1 | - | 5 | 425 | |||
CR749519.1 | - | 6 | 569 |