Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138477.2 | NP_612486.2 | 6 | 1191 | ||
XM_005254176.4 | XP_005254233.1 | 6 | 1106 | |||
XM_011521270.2 | XP_011519572.1 | 6 | 1131 | |||
XM_011521271.2 | XP_011519573.1 | 6 | 1128 | |||
XM_011521274.2 | XP_011519576.1 | 3 | 177 | |||
GenBank mRNA | AF525398.1 | - | 6 | 1188 | ||
BC052568.1 | - | 6 | 1079 | |||
BC066640.1 | - | 6 | 1079 | 6577335 |