Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013838.1 | NP_001013860.1 | 14 | 1214 | ||
NM_001317026.1 | NP_001303955.1 | 14 | 1214 | |||
XM_011522874.1 | XP_011521176.1 | 14 | 1352 | |||
XM_011522875.2 | XP_011521177.2 | 14 | 1352 | |||
XM_017022953.1 | XP_016878442.1 | 14 | 1352 | |||
GenBank mRNA | AB113647.1 | - | 14 | 1214 | ||
AK090421.1 | - | 2 | 113 | |||
AL832446.1 | - | 13 | 1066 | |||
FJ026014.1 | - | 14 | 1094 |