Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199317.1 | NP_001186246.1 | 1 | 376 | ||
NM_144673.2 | NP_653274.1 | 1 | 376 | |||
GenBank mRNA | AF479260.1 | - | 1 | 368 | ||
AK098598.1 | - | 1 | 361 | |||
AY862139.1 | - | 1 | 376 | |||
BC025354.1 | - | 1 | 367 | 4904062 | ||
BI830241.1 | - | 1 | 376 |