Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144674.1 | NP_653275.1 | 3 | 760 | ||
XM_011522383.2 | XP_011520685.1 | 3 | 760 | |||
XM_011522385.2 | XP_011520687.1 | 3 | 760 | |||
XM_011522389.1 | XP_011520691.1 | 3 | 760 | |||
XM_011522390.2 | XP_011520692.1 | 6 | 1117 | |||
XM_017022955.1 | XP_016878444.1 | 3 | 760 | |||
XM_017022956.1 | XP_016878445.1 | 3 | 657 | |||
XM_017022957.1 | XP_016878446.1 | 3 | 760 | |||
GenBank mRNA | AK057433.1 | - | 3 | 760 | ||
BC130336.1 | - | 3 | 743 | 40146367 | ||
BC130338.1 | - | 3 | 743 | 40146369 |