Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153350.3 | NP_699181.2 | 2 | 896 | ||
GenBank mRNA | BC036680.2 | - | 3 | 771 | 5262152 |