Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109659.1 | NP_001103129.1 | 2 | 143 | ||
NM_001109660.1 | NP_001103130.1 | 2 | 223 | |||
GenBank mRNA | AK058133.1 | - | 2 | 223 | ||
BC101433.1 | - | 2 | 143 | 40021839 | ||
BC101434.1 | - | 2 | 143 | 40021838 | ||
BC101435.1 | - | 2 | 143 | 40021831 | ||
BC101436.1 | - | 2 | 223 | 40021833 | ||
BC127955.1 | - | 2 | 223 | 40023544 |