Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080556.1 | NP_001074025.1 | 9 | 1052 | ||
NM_145054.4 | NP_659491.4 | 10 | 1256 | |||
XM_017024227.1 | XP_016879716.1 | 9 | 1041 | |||
GenBank mRNA | AB065281.1 | - | 10 | 1221 | ||
AK074435.1 | - | 9 | 1036 | |||
AK094847.1 | - | 11 | 1403 | |||
AK315679.1 | - | 10 | 1211 | |||
BC025392.2 | - | 10 | 1216 | 4838276 |