Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033576.1 | NP_001028748.1 | 5 | 506 | ||
NM_001267052.1 | NP_001253981.1 | 5 | 503 | |||
NM_001308281.1 | NP_001295210.1 | 4 | 409 | |||
NM_173167.3 | NP_775259.1 | 4 | 409 | |||
XM_017024234.1 | XP_016879723.1 | 5 | 527 | |||
GenBank mRNA | AF539794.1 | - | 4 | 409 | ||
AL832355.1 | - | 5 | 500 | |||
AL833281.1 | - | 5 | 479 | |||
BC101062.2 | - | 4 | 409 | 40008186 | ||
BC101063.2 | - | 4 | 409 | 40008187 | ||
BX647642.1 | - | 5 | 506 |