Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166131.1 | NP_001159603.1 | 2 | 395 | ||
NM_152463.2 | NP_689676.2 | 2 | 395 | |||
XM_017024236.1 | XP_016879725.1 | 2 | 395 | |||
GenBank mRNA | AK055926.1 | - | 2 | 386 | ||
BC016470.2 | - | 1 | 338 | 2899969 | ||
DA006645.1 | - | 2 | 390 | |||
DB245967.1 | - | 2 | 396 |