Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055926.1 | - | 2 | 284 | ||
AK308275.1 | - | 2 | 259 | |||
BC016470.2 | - | 1 | 236 | 2899969 |