Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 1, 2 | 293 | ||
GenBank mRNA | AK223584.1 | - | 1, 2 | 227 | ||
AK291475.1 | - | 1, 2 | 216 | |||
AK311363.1 | - | 175 | ||||
AY074907.1 | - | 1, 2 | 203 | |||
BC033113.1 | - | 1, 2 | 210 | 5429060 | ||
DC327155.1 | - | 1, 2 | 293 |