Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304949.1 | NP_001291878.1 | 5 | 1174 | ||
NM_001304951.1 | NP_001291880.1 | 2 | 863 | |||
NM_001304952.1 | NP_001291881.1 | 5 | 1095 | |||
NM_001304953.1 | NP_001291882.1 | 5 | 1393 | |||
NM_001304954.1 | NP_001291883.1 | 4 | 1192 | |||
NM_152465.2 | NP_689678.1 | 4 | 1128 | |||
XM_005257927.4 | XP_005257984.1 | 5 | 1364 | |||
XM_006721723.3 | XP_006721786.1 | 5 | 1361 | |||
XM_011524398.2 | XP_011522700.1 | 5 | 1094 | |||
XM_011524399.2 | XP_011522701.1 | 4 | 1009 | |||
XM_011524400.2 | XP_011522702.1 | 4 | 918 | |||
XM_011524401.1 | XP_011522703.1 | 3 | 919 | |||
XM_017024237.1 | XP_016879726.1 | 4 | 1012 | |||
GenBank mRNA | AK301868.1 | - | 5 | 1092 | ||
BC029574.1 | - | 4 | 1075 | 5267334 | ||
BQ215318.1 | - | 2 | 703 |