Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152468.4 | NP_689681.2 | 8 | 1346 | ||
XM_011524402.2 | XP_011522704.1 | 8 | 2836 | |||
XM_011524403.2 | XP_011522705.1 | 8 | 2823 | |||
XM_011524404.2 | XP_011522706.1 | 8 | 2835 | |||
XM_011524406.2 | XP_011522708.1 | 8 | 2838 | |||
XM_011524409.2 | XP_011522711.1 | 8 | 2840 | |||
XM_011524410.2 | XP_011522712.1 | 8 | 2838 | |||
XM_011524411.2 | XP_011522713.1 | 8 | 2838 | |||
XM_017024238.1 | XP_016879727.1 | 8 | 2837 | |||
XM_017024239.1 | XP_016879728.1 | 8 | 2837 | |||
XM_017024240.1 | XP_016879729.1 | 8 | 2837 | |||
XM_017024241.1 | XP_016879730.1 | 8 | 2836 | |||
XM_017024242.1 | XP_016879731.1 | 2 | 226 | |||
XM_017024243.1 | XP_016879732.1 | 5 | 547 | |||
XM_017024244.1 | XP_016879733.1 | 8 | 2826 | |||
GenBank mRNA | AK090478.1 | - | 8 | 1306 | ||
AK303499.1 | - | 7 | 1094 | |||
AY057380.1 | - | 7 | 964 | |||
AY099358.1 | - | 8 | 1336 | |||
AY099359.1 | - | 7 | 855 | |||
AY236500.1 | - | 7 | 1033 | |||
BC110296.1 | - | 8 | 1338 | 5211834 | ||
BC141865.1 | - | 7 | 1190 | 40075995 |