Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004428.2 | NP_004419.2 | 5 | 627 | ||
NM_182685.1 | NP_872626.1 | 4 | 561 | |||
GenBank mRNA | AK057845.1 | - | 5 | 1040 | ||
AK289589.1 | - | 4 | 558 | |||
AK313117.1 | - | 5 | 671 | |||
AL833641.1 | - | 4 | 559 | |||
BC032698.1 | - | 5 | 620 | 5402648 | ||
BC095432.1 | - | 5 | 609 | 30376031 | ||
BC150578.1 | - | 2 | 425 | |||
CR457416.1 | - | 5 | 522 | |||
M57730.1 | - | 5 | 595 |