Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133264.4 | NP_573571.1 | 3 | 410 | ||
XM_005257083.2 | XP_005257140.1 | 3 | 412 | |||
XM_005257084.2 | XP_005257141.1 | 3 | 370 | |||
XM_011524412.1 | XP_011522714.1 | 3 | 428 | |||
GenBank mRNA | AB043786.1 | - | 2 | 170 | ||
AF370396.1 | - | 3 | 334 | |||
AJ431177.1 | - | 2 | 170 | |||
AK055057.1 | - | 3 | 401 | |||
AK289578.1 | - | 3 | 370 | |||
AK289779.1 | - | 3 | 429 | |||
AK290301.1 | - | 3 | 406 | |||
AK311544.1 | - | 3 | 428 | |||
AK315836.1 | - | 3 | 390 | |||
AL834239.1 | - | 2 | 279 | |||
BC065551.1 | - | 3 | 347 | 6044222 | ||
CN404607.1 | - | 3 | 410 |