Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195386.1 | NP_001182315.1 | 3 | 1014 | ||
NM_001195387.1 | NP_001182316.1 | 3 | 998 | |||
NM_145274.3 | NP_660317.2 | 3 | 1006 | |||
XM_005257089.4 | XP_005257146.1 | 4 | 1203 | |||
XM_017024253.1 | XP_016879742.1 | 4 | 1171 | |||
GenBank mRNA | AK298616.1 | - | 3 | 949 |