Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008239.2 | NP_001008240.1 | 2 | 446 | ||
NM_145055.3 | NP_659492.1 | 2 | 446 | |||
XM_011525822.1 | XP_011524124.1 | 2 | 297 | |||
XM_017025555.1 | XP_016881044.1 | 2 | 293 | |||
GenBank mRNA | AK289738.1 | - | 2 | 333 | ||
AK292981.1 | - | 2 | 434 | |||
BC016149.1 | - | 1 | 119 | 4040087 | ||
BC084579.1 | - | 2 | 413 | 6141272 | ||
BU192741.1 | - | 2 | 411 | |||
BX640683.1 | - | 1 | 127 | |||
DB284444.1 | - | 2 | 448 |