Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308238.1 | NP_001295167.1 | 9 | 904 | ||
NM_173505.3 | NP_775776.2 | 10 | 1003 | |||
XM_011525830.2 | XP_011524132.1 | 9 | 923 | |||
XM_011525831.2 | XP_011524133.1 | 9 | 2943 | |||
XM_011525832.2 | XP_011524134.1 | 9 | 3061 | |||
XM_017025561.1 | XP_016881050.1 | 8 | 827 | |||
GenBank mRNA | AK057782.1 | - | 9 | 834 | ||
AK123197.1 | - | 7 | 2323 | |||
AK313667.1 | - | 10 | 921 | |||
BC030622.1 | - | 10 | 1008 | 4818619 |