Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153000.4 | NP_694545.1 | 5 | 1628 | ||
GenBank mRNA | AB056722.1 | - | 5 | 1627 | ||
AB104887.1 | - | 5 | 1627 | |||
AF461902.1 | - | 4 | 1224 | |||
AK300743.1 | - | 6 | 1430 | |||
AK303076.1 | - | 4 | 1220 | |||
BC053324.1 | - | 6 | 1599 | 6644230 |