Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153000.4 | NP_694545.1 | 3 | 621 | ||
XM_011525603.1 | XP_011523905.1 | 3 | 614 | |||
GenBank mRNA | AB056722.1 | - | 3 | 620 | ||
AB104887.1 | - | 3 | 620 | |||
AF461902.1 | - | 2 | 218 | |||
AK300743.1 | - | 4 | 423 | |||
AK303076.1 | - | 2 | 213 | |||
BC053324.1 | - | 4 | 592 | 6644230 |