Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297624.1 | NP_001284553.1 | 4 | 331 | ||
NM_001297625.1 | NP_001284554.1 | 4 | 333 | |||
NM_144684.3 | NP_653285.2 | 5 | 460 | |||
XM_011526465.2 | XP_011524767.1 | 5 | 862 | |||
GenBank mRNA | AK096442.1 | - | 3 | 196 | ||
AK096559.1 | - | 5 | 450 | |||
AK303271.1 | - | 4 | 322 | |||
AL832560.1 | - | 4 | 310 | |||
AY512662.1 | - | 5 | 429 | |||
BC007056.1 | - | 5 | 455 | |||
BC018783.1 | - | 5 | 429 | 4873910 | ||
BC054018.1 | - | 5 | 460 | |||
BC065503.1 | - | 5 | 435 | |||
DA501214.1 | - | 5 | 458 |