Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144685.4 | NP_653286.2 | 3 | 1976 | ||
XM_006723036.2 | XP_006723099.1 | 3 | 1951 | |||
GenBank mRNA | AK057380.1 | - | 3 | 1726 | ||
AK292228.1 | - | 3 | 1937 | |||
AK301713.1 | - | 4 | 1729 | |||
AK302301.1 | - | 3 | 1333 | |||
BC034501.2 | - | 3 | 1942 | 4823304 |