Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145303.2 | NP_001138775.2 | 8 | 1415 | ||
NM_144686.3 | NP_653287.2 | 8 | 1397 | |||
GenBank mRNA | AK314973.1 | - | 8 | 1318 | ||
AY236492.1 | - | 8 | 1414 | |||
AY263166.1 | - | 8 | 1294 | |||
BC025323.1 | - | 8 | 1304 | 2959382 | ||
BC032477.1 | - | 2 | 434 |