Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005227.2 | NP_005218.1 | 2 | 474 | ||
NM_182689.1 | NP_872631.1 | 2 | 474 | |||
NM_182690.2 | NP_872632.2 | 2 | 474 | |||
GenBank mRNA | AJ006352.1 | - | 2 | 414 | ||
AJ006353.1 | - | 2 | 414 | |||
BC107483.1 | - | 2 | 464 | 40030599 | ||
BE780161.1 | - | 1 | 28 | |||
BM740848.1 | - | 2 | 474 | |||
BM747625.1 | - | 2 | 474 | |||
CD671399.1 | - | 2 | 480 | |||
CR533569.1 | - | 2 | 387 | |||
U14188.1 | - | 2 | 414 |