Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145276.2 | NP_660319.1 | 4 | 1033 | ||
XM_005259750.4 | XP_005259807.1 | 4 | 510 | |||
XM_005259751.3 | XP_005259808.1 | 5 | 915 | |||
XM_006722650.3 | XP_006722713.1 | 3 | 970 | |||
XM_006722651.3 | XP_006722714.1 | 4 | 863 | |||
XM_011527698.1 | XP_011526000.1 | 4 | 853 | |||
XM_011527699.2 | XP_011526001.1 | 3 | 1037 | |||
XM_011527700.2 | XP_011526002.1 | 3 | 808 | |||
XM_017026332.1 | XP_016881821.1 | 4 | 651 | |||
GenBank mRNA | AK092116.1 | - | 5 | 915 | ||
AK313755.1 | - | 4 | 937 | |||
BC022523.1 | - | 4 | 1001 | 4794529 |