Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301046.1 | NP_001287975.1 | 3 | 2499 | ||
NM_145056.2 | NP_659493.2 | 4 | 2807 | |||
XM_011526499.2 | XP_011524801.1 | 4 | 3583 | |||
XM_011526500.2 | XP_011524802.1 | 3 | 2499 | |||
XM_017026334.1 | XP_016881823.1 | 4 | 2668 | |||
GenBank mRNA | BC016161.1 | - | 3 | 2498 | 2967514 |