Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723038.3 | XP_006723101.1 | 5 | 612 | ||
XM_006723039.3 | XP_006723102.1 | 4 | 662 | |||
XM_011526503.2 | XP_011524805.1 | 5 | 722 | |||
XM_011526504.2 | XP_011524806.1 | 4 | 536 | |||
XM_011526505.2 | XP_011524807.1 | 6 | 694 | |||
XM_011526507.2 | XP_011524809.1 | 3 | 370 | |||
XM_011526508.2 | XP_011524810.1 | 2 | 328 | |||
XM_011526510.2 | XP_011524812.1 | 3 | 507 | |||
XM_011526511.2 | XP_011524813.1 | 5 | 612 | |||
XM_017026336.1 | XP_016881825.1 | 4 | 536 | |||
XM_017026337.1 | XP_016881826.1 | 5 | 627 | |||
XM_017026338.1 | XP_016881827.1 | 2 | 282 | |||
XM_017026339.1 | XP_016881828.1 | 6 | 694 | |||
XM_017026340.1 | XP_016881829.1 | 6 | 694 | |||
GenBank mRNA | AK056753.1 | - | 5 | 591 | ||
AK315728.1 | - | 5 | 607 | |||
BC026210.2 | - | 5 | 546 | 3028503 |