Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134444.4 | NP_604393.2 | 9 | 3134 | ||
XM_017026344.1 | XP_016881833.1 | 7 | 2609 | |||
XM_017026345.1 | XP_016881834.1 | 7 | 2606 | |||
GenBank mRNA | AF442488.1 | - | 8 | 2769 | ||
AF479747.1 | - | 8 | 2781 | |||
AF482706.1 | - | 8 | 2779 | |||
AK056688.1 | - | 7 | 2661 | |||
AK315003.1 | - | 9 | 3134 | |||
AY072792.1 | - | 8 | 2772 | |||
BC016443.1 | - | 6 | 1241 | |||
BC050326.1 | - | 9 | 2808 | 4837670 |